Learn More
Apexbio Technology LLC U-104 178606-66-1 25mg
SDP
Supplier:  Apexbio Technology LLC A435825
Encompass_Preferred
U-104 (CAS 178606-66-1) is a urea-based sulfonamide that functions as an inhibitor of carbonic anhydrase IX (CAIX) an enzyme upregulated in hypoxic tumor microenvironments In preclinical studies U-104 suppressed primary tumor growth in murine models compared to vehicle-treated controls Additionally in the 4T1 experimental metastasis model administration of U-104 reduced the formation of metastatic lesions These findings underscore the utility of U-104 as a tool compound for investigating CAIX-mediated tumor progression and its role in tumor hypoxia growth and metastasis
Catalog No. 50-000-96118
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.