Learn More
Eurofins Discoverx U-18666A, 5 mg. (92-1094)
Supplier:  Eurofins Discoverx 921094
Encompass
U 18666A is a small molecule inhibitor of cholesterol transport from endosomes/lysosomes to the endoplasmic reticulum in a variety of cell types, including macrophages and sympathetic neurons. Also weakly inhibits hedgehog signaling. Implicated in multiple diseases including Neimann-Pick disease type C (NPC), Alzheimer's, cataracts, and atherosclerosis. Provided as 5 mg of dry powder. These highly purified, pre-validated control ligands have been used to qualify our InCELL Hunter,  PathHunter and cAMP Hunter cell lines as well as our Assay Ready Kits. Ligands are conveniently supplied with a recommended reconstitution buffer. For detailed information and product-specific datasheet, visit www.discoverx.com/ligands.
Catalog No. 50-199-9632
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.