Learn More
Enzo Life Sciences U-69593 (5mg). CAS: 96744-75-1
SDP
Supplier:  Enzo Life Sciences BMLN7050005
Encompass_Preferred
Selective κ-opioid agonist. Thought to be selective for the κ-1 subtype (KD=3.8nM in human cerebral cortex membranes). Immunomodulatory activity. Purity: ≥99% (TLC). Solubility: Soluble in 100% ethanol, 0.1N hydrogen chloride, or DMSO. Insoluble in water or 0.1N sodium hydroxide. Long Term Storage: -20°C.
Catalog No. 50-201-1175
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.