Learn More
Abcam U-73343, Inactive PLC Inhibitor Analog, Solid, 25 mg
Description
- Molecular weight: 466.7 Da
- Purity: >98%
- Function: Negative control for phospholipase C (PLC) inhibitor U73122 (ab120998)
- Physical form: Solid
- Chemical composition: 1-[6-((17b-3-Methoxyestra-1,3,5(10)-trien-17-yl)amino)hexyl]-2,5-pyrrolidinedione
- CAS number: 142878-12-4
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Target Molecule | Phospholipase C |
| Shipping Condition | Ambient |
| Form | Solid |
| Buffer | 100% 1-[6-((17b-3-Methoxyestra-1,3,5(10)-trien-17-yl)amino)hexyl]-2,5-pyrrolidinedione |
| Content And Storage | Store at ambient temperature. |
| Quantity | 25 mg |
| For Use With (Application) | Negative control for phospholipase C (PLC) inhibitor U73122 |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.