Learn More
Hunt Optics And Imaging U PLAN FLUORITE 4X PHASE IPC
SDP
Supplier:  Hunt Optics And Imaging 1U2PC5222
Encompass
UPLFLN4XIPC-2; U PLAN FLUORITE 4X PHASE IPC OBJ, NA 0.13, WD 16.4MM
Catalog No. NC2600560