Learn More
Montana Molecular Red Upward DAG Sensor Kit U0300R, 30mL, CMV
SDP
Supplier:  Montana Molecular U0300R030
Encompass_Preferred
Measure diacylglycerol (DAG) signaling kinetics, screen GPCRs for Gq activity, and more with this live cell fluorescent assay. Add the DAG sensor to your cells and measure robust signals after 30 hours on an automated plate reader or fluorescence imaging system. Increasing red fluorescence indicates DAG increase. These assays are genetically encoded and packaged in BacMam for efficient transduction in most cell types - including uniform and near 100% efficiency in HEK293 cells as well as expression in primary or iPSC-derived cells. Kits include positive control receptors and agonists.
Catalog No. 50-163-1925
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.