Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

U2AF1L4 Monoclonal Antibody (OTI8B1), TrueMAB™, OriGene
SDP

Catalog No. 501683390
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-168-3390 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-168-3390 Supplier OriGene Supplier No. TA810435
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

RNA-binding protein that function as a pre-mRNA splicing factor. Plays a critical role in both constitutive and enhancer-dependent splicing by mediating protein-protein interactions and protein-RNA interactions required for accurate 3′-splice site selection. Acts by enhancing the binding of U2AF2 to weak pyrimidine tracts. Also participates in the regulation of alternative pre-mRNA splicing. Activates exon 5 skipping of PTPRC during T cell activation; an event reversed by GFI1. Binds to RNA at the AG dinucleotide at the 3′-splice site.

Specifications

Antigen U2AF1L4
Applications Western Blot
Classification Monoclonal
Clone OTI8B1
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene U2AF1L4
Gene Accession No. Q8WU68
Gene Alias U2AF1-RS3, U2AF1L3, U2AF1L3V1, U2AF1RS3, U2af26
Gene Symbols U2AF1L4
Host Species Mouse
Immunogen Full length human recombit protein of human U2AF1L4 produced in E.coli.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 199746
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.