Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Ub-K63 Monoclonal Antibody (HWA4C4), eBioscience™, Invitrogen™
GREENER_CHOICE

Catalog No. 5013454
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-134-54 100 μg
50-134-53 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-134-54 Supplier Invitrogen™ Supplier No. 14607782
  • $467.00
    $350.65 / Each of 1

    Save $116.35

    25% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Mouse Monoclonal Antibody

Description: Ubiquitin is a highly conserved 76kDa protein which is ligated to many proteins in both monomeric and polymeric forms. Ubiquitin polymers (polyubiquitin) can be found in several varieties depending on which of its 7 lysine residues is the site of ligation. The two most common polymer chains are linked via an isopeptide bond between glycine-76 and either lysine-48 (K-48) or lysine-63 (K-63). K-48-linked polyubiquitination typically results in targeting the protein to the 26S proteosome resulting in proteolysis, although recently exceptions to this have been observed. In contrast, K-63-linked polyubiquitination results in signals related to intracellular trafficking, cell signaling, ribosomal biogenesis, and DNA damage repair. There are also reports that K63-linked polyubiquitin is involved in proteosome-independent proteolysis via autophagy. HWA4C4 reacts specifically with the isopeptide linkage region of lysine-63-linked polyubiquitin. It does not react with other forms of ubiquitin including ubiquitin monomer and lysine-48-linked chains. Applications Reported: This HWA4C4 antibody has been reported for use in immunoblotting (WB). Applications Tested: This HWA2-8°C4 antibody has been tested by immunoblot of lysine-63-linked polyubiquitin chains. This can be used at less than or equal to 2 μg/mL. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.

Ubiquitin is a conserved 76 amino acid polypeptide and can affect proteasomal degradation of the protein it is bound to, or mediate interactions with other proteins related to post-translational modifications. The degradation of cellular regulatory proteins by the Uubiquitin pathway is important as it controls the cellular growth and proliferation. Ubiquitin-dependent proteolysis occurs after a covalent attachment of the peptide to a lysine residue of a protein, which involves three enzymatic reactions: E1, E2 and E3. The first reaction involves ubiquitin-activating enzyme. The third reaction uses enzyme ubiquitin ligase (E3) to transfer the activated ubiquitin from E2 to a lysine residue on a protein, or directly transfers the ubiquitin from E2 to the substrate.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Ub-K63
Applications Western Blot
Classification Monoclonal
Clone HWA4C4
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.2
Gene UBB
Gene Accession No. P0CG47, P0CG48, P62979, P62987
Gene Alias 0610006J14Rik; 2700054O04Rik; 40S ribosomal protein S27a; 60S ribosomal protein L40; AI194771; AL033289; CEP52; CEP80; D8Ertd21e; epididymis luminal protein 112; epididymis secretory protein Li 50; Gm1863; HEL112; HEL-S-50; HMG20; HUBCEP52; L40; Large ribosomal subunit protein eL40; LOC100912032; polyubiquitin B; polyubiquitin C; polyubiquitin-B; Polyubiquitin-C; ribosomal protein S27A; RPL40; Rps27a; S27A; Small ribosomal subunit protein eS31; TI-225; Uba52; UBA80; Ubb; Ubb2; UBC; UBCEP1; UBCEP2; UBCEP80; Ubiquitin; Ubiquitin A-52 residue ribosomal protein fusion product 1; ubiquitin and ribosomal protein S27a; ubiquitin B; ubiquitin C; ubiquitin carboxyl extension protein 52; ubiquitin carboxyl extension protein 80; ubiquitin/60S ribosomal fusion protein; Ubiquitin-40S ribosomal protein S27a; ubiquitin-40S ribosomal protein S27a-like; ubiquitin-52 amino acid fusion protein; ubiquitin-60S ribosomal protein L40; ubiquitin-CEP52; ubiquitin-CEP80; Ubiquitin-related; Ubiquitin-related 1; Ubiquitin-related 2
Gene Symbols RPS27A, UBA52, UBB, UBC
Host Species Mouse
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6233, 7311, 7314, 7316
Target Species Human
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.