Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

UBASH3A Monoclonal Antibody (OTI4H1), TrueMAB™, OriGene
SDP

Catalog No. 501682655
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-168-2655 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-168-2655 Supplier OriGene Supplier No. TA809237
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

TULA (T cell ubiquitin ligand, UBASH3A, ubiquitin associated and SH3 domain containing A), STS-2 or CLIP4, is a 661 amino acid protein that localizes to both the nucleus and the cytoplasm and contains one SH3 domain and one UBA domain. Expressed at high levels in thymus, bone marrow, spleen and peripheral blood leukocytes, TULA exists as either a homodimer or a homo-oligomer that interferes with the degradation of receptor-type tyrosine kinases and promotes the accumulation of activated receptors on the cell surface. Additionally, TULA is part of an EGFR- and Cbl-containing complex that interacts with ubiquitinated proteins. The gene encoding TULA, which maps to human chromosome 21, may be involved in the pathogenesis of type I diabetes. Multiple isoforms of TULA are produced due to alternative splicing events.

Specifications

Antigen UBASH3A
Applications Western Blot
Classification Monoclonal
Clone OTI4H1
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene UBASH3A
Gene Accession No. P57075
Gene Alias CLIP4, STS-2, TULA, TULA-1
Gene Symbols UBASH3A
Host Species Mouse
Immunogen Human recombit protein fragment corresponding to amino acids 288-549 of human UBASH3A produced in E.coli.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 53347
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.