Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ UBE1 Isoform A/B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA517274
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA517274 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA517274 Supplier Invitrogen™ Supplier No. PA517274
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

It is not recommended to aliquot this antibody.

UBE1 catalyzes the first step in ubiquitin conjugation to mark cellular proteins for degradation. This gene complements an X-linked mouse temperature-sensitive defect in DNA synthesis, and thus may function in DNA repair. It is part of a gene cluster on chromosome Xp11. 23. Alternative splicing results in 2 transcript variants encoding the same protein, but with different 5' UTR.
TRUSTED_SUSTAINABILITY

Specifications

Antigen UBE1 Isoform A/B
Applications Western Blot
Classification Polyclonal
Concentration 20 μg/mL
Conjugate Unconjugated
Formulation 0.01M HEPES with 100μg/mL BSA, 50% glycerol, 0.15M NaCl and no preservative; pH 7.5
Gene UBA1
Gene Accession No. P22314, Q02053, Q5U300
Gene Alias A1S9; A1S9T; A1S9T and BN75 temperature sensitivity complementing; A1ST; AMCX1; CFAP124; EMBL:AAH85791.1, ECO:0000312; GXP1; POC20; POC20 centriolar protein homolog; Protein A1S9; RGD:1359327}; Sbx; SMAX2; testicular secretory protein Li 63; UBA1; uba1 {ECO:0000312; UBA1, ubiquitin-activating enzyme E1 homolog A; UBA1A; UBE1; Ube-1; Ube1ax; Ube1x; ubiquitin like modifier activating enzyme 1; Ubiquitin-activating enzyme E1; ubiquitin-activating enzyme E1 {ECO:0000250; Ubiquitin-activating enzyme E1 X; ubiquitin-activating enzyme E1, Chr X; ubiquitin-like modifier activating enzyme 1; ubiquitin-like modifier-activating enzyme 1; ubiquitin-like modifier-activating enzyme 1 {ECO:0000250; Ubiquitin-like modifier-activating enzyme 1 X; UniProtKB:Q02053}
Gene Symbols UBA1
Host Species Rabbit
Immunogen Synthetic peptide surrounding Thr800 of human UBE1.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22201, 314432, 7317
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.