Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ UBE2D2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA552024
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA552024 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA552024 Supplier Invitrogen™ Supplier No. PA552024
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LTDLQRDPPA QCSAGPVGDD LFHWQATIMG PNDSPYQGGV FFLTIHFPTD YPFKPPKVAF TTKIYHPNIN SNGSICLDIL RS Highest antigen sequence identity to the following orthologs: Mouse - 95%, Rat - 95%.

The modification of proteins with ubiquitin is an important cellular mechanism for targeting abnormal or short-lived proteins for degradation. Ubiquitination involves at least three classes of enzymes: ubiquitin-activating enzymes, or E1s, ubiquitin-conjugating enzymes, or E2s, and ubiquitin-protein ligases, or E3s. This gene encodes a member of the E2 ubiquitin-conjugating enzyme family. This enzyme functions in the ubiquitination of the tumor-suppressor protein p53, which is induced by an E3 ubiquitin-protein ligase. Two alternatively spliced transcript variants have been found for this gene and they encode distinct isoforms.
TRUSTED_SUSTAINABILITY

Specifications

Antigen UBE2D2
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.10 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene UBE2D2
Gene Accession No. P62837, P62838, P62839
Gene Alias (E3-independent) E2 ubiquitin-conjugating enzyme D2; 1500034D03Rik; E2 ubiquitin conjugating enzyme UbcH5B; E2 ubiquitin-conjugating enzyme D2; E2(17)KB 2; E2(17)KB2; E217kB; p53-regulated ubiquitin-conjugating enzyme 1; PUBC1; similar to ubiquitin-conjugating enzyme E2D 2; Ubc2e; ubc2e ubiquitin conjugating enzyme; ubc4; UBC4/5; UBC5B; UBCH4; UBCH5B; ube2d2; Ube2d2a; Ubiquitin carrier protein D2; ubiquitin conjugating enzyme 2e; ubiquitin conjugating enzyme E2 D2; ubiquitin conjugating enzyme E2D; ubiquitin conjugating enzyme E2D 2; ubiquitin-conjugating enzyme E2 D2; Ubiquitin-conjugating enzyme E2(17)KB 2; ubiquitin-conjugating enzyme E2-17 kDa 2; ubiquitin-conjugating enzyme E2D 2; ubiquitin-conjugating enzyme E2D 2 (homologous to yeast UBC4/5); ubiquitin-conjugating enzyme E2D 2 (UBC4/5 homolog, yeast); ubiquitin-conjugating enzyme E2D 2A; ubiquitin-protein ligase D2; Unknown (protein for MGC:138112); zgc:55886; zgc:55886 protein; zgc:77149; zmp:0000000618
Gene Symbols UBE2D2, Ube2d2a
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human UBE2D4.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 56550, 641452, 7322
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.