Learn More
Invitrogen™ UBE2L3 Polyclonal Antibody

Description
This antibody is predicted to react with bovine based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 128,000.
Specifications
Specifications
| Antigen | UBE2L3 |
| Applications | Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 |
| Gene | UBE2L3 |
| Gene Accession No. | P68036, P68037 |
| Gene Alias | C79827; E2 ubiquitin-conjugating enzyme L3; E2-F1; L-UBC; UBCE7; UBCH7; UbcM4; Ube2l3; Ubiquitin carrier protein L3; ubiquitin conjugating enzyme E2 L3; ubiquitin conjugating enzyme E2L 3; ubiquitin-conjugating enzyme 7; ubiquitin-conjugating enzyme E2 L3; Ubiquitin-conjugating enzyme E2-F1; ubiquitin-conjugating enzyme E2L 3; ubiquitin-conjugating enzyme UBCH7; ubiquitin-protein ligase L3; Unknown (protein for MGC:127956) |
| Gene Symbols | UBE2L3 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.