Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ UBE2V1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA592896
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA592896 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA592896 Supplier Invitrogen™ Supplier No. PA592896
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MAATTGSGVK VPRNFRLLEE LEEGQKGVGD GTVSWGLEDD EDMTLTRWTG MIIGPPRTIY ENRIYSLKIE CGPKYPEAPP FVRFVTKINM NGVNSSNGVV DPRAISVLAK WQNSYSIKVV LQELRRLMMS KENMKLPQPP EGQCYSN; Positive Samples: BT-474, 293T, HepG2, HeLa, Mouse brain, Mouse pancreas, Mouse spinal cord, Rat liver, Rat spleen; Cellular Location: Nucleus.

The TMEM189-UEV mRNA is an infrequent but naturally occurring co-transcribed product of the neighboring TMEM189 and UBE2V1 genes. Ubiquitin-conjugating E2 enzyme variant proteins constitute a distinct subfamily within the E2 protein family. They have sequence similarity to other ubiquitin-conjugating enzymes but lack the conserved cysteine residue that is critical for the catalytic activity of E2s. The protein produced by this transcript has UEV1 B domains but the protein is localized to the cytoplasm rather than to the nucleus. The significance of this co-transcribed mRNA and the function of its protein product has not yet been determined.
TRUSTED_SUSTAINABILITY

Specifications

Antigen UBE2V1
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.33 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene Ube2v1
Gene Accession No. Q13404, Q9CZY3
Gene Alias 0610011J09Rik; AI256840; CIR1; CROC1; CROC-1; CROC1B; CROC-1B; CROC-1UBE2V; D7Bwg1382e; DJ1185N5.1.3; DNA-binding protein; KUA-UEV; P/OKcl.19; TRAF6-regulated IKK activator 1 beta Uev1A; UBE2V; Ube2v1; ubiquitin conjugating enzyme E2 V1; ubiquitin conjugating enzyme E2 variant 1; ubiquitin-conjugating enzyme E2 variant 1; UEV1; UEV-1; UEV1A; UEV-1CIR1
Gene Symbols Ube2v1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-147 of human UBE2V1 (NP_0010274591).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 296390, 66589, 7335
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.