Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Ubiquitin B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA595195
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA595195 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA595195 Supplier Invitrogen™ Supplier No. PA595195
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: human Caco-2 whole cell, human K562 whole cell, human THP-1 whole cell, rat PC-12 whole cell, rat brain tissue, mouse brain tissue. IHC: rat intestine tissue, human mammary cancer tissue, mouse intestine tissue. ICC/IF: A431 cell. Store at -20°C for one year from date of receipt. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for six months. Avoid repeated freeze-thaw cycles.

Ubiquitin is a highly conserved protein of about 8.5 kDa molecular weight, which has an ATP dependent role in the targeting of proteins for proteolytic degradation. To perform this function, the protein to be degraded is first covalently attached to the C terminus of ubiquitin, and the ubiquitinated complex is then recognized by a complex of degradative enzymes. Interestingly, ubiquitin also becomes covalently bonded to many types of pathological inclusions, which appear to be resistant to normal degradation. Therefore, ubiquitin antibodies are very useful for studies of these inclusions. For example, the neurofibrillary tangles and paired helical filaments diagnostic of Alzheimer's disease, Lewy bodies seen in Parkinson's disease, and Pick bodies found in Pick's disease are all heavily ubiquitinated and can be readily visualized with ubiquitin antibodies.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Ubiquitin B
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 500 μg/mL
Conjugate Unconjugated
Formulation PBS with 5mg BSA and 0.05mg sodium azide
Gene UBB
Gene Accession No. P0CG47, P0CG49, P0CG51
Gene Alias AL033289; epididymis secretory protein Li 50; HEL-S-50; HUBCEP52; im:6892314; im:6892314 protein; MGC127041; polyubiquitin; polyubiquitin B; polyubiquitin-B; RPL40; Rps27a; si:ch211-202a12.3; Uba52; Ubb; Ubb2; UBC; Ubi-p63E; Ubiquitin; ubiquitin B; ubiquitin C; Ubiquitin-related; zgc:172187; zUBC
Gene Symbols UBB
Host Species Rabbit
Immunogen E.coli-derived human Ubiquitin recombinant protein (Position: M77-G152).
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 192255, 22187, 7314
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.