Learn More
Enzo Life Sciences Ubiquitin (human) (recombinant) (5 mg) CAS: 79586-22-4
SDP
Supplier:  Enzo Life Sciences BMLUW87950005
Encompass_Preferred
MW: 8565 Da. Purity: ≥98% (SDS-PAGE). Formulation: Lyophilized. Appearance: White powder. Source: Produced in E. coli. Solubility: Soluble in water UniProt: P0CG47 (UBB), P0CG48 (UBC), P62979 (RPS27A), P62987 (UBA52). Long Term Storage: -20°C
Catalog No. NC0751403
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.