Learn More
Invitrogen™ UBR2 Monoclonal Antibody (2G10)

Description
Adding 0.2 mL of distilled water will yield a concentration of 500 μg/mL. Positive Control - WB: human HEK293 whole cell, human Jurkat whole cell, human K562 whole cell. Store at -20°C for one year from date of receipt. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for six months. Avoid repeated freeze-thaw cycles.
Specifications
Specifications
| Antigen | UBR2 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Clone | 2G10 |
| Concentration | 500 μg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 4mg trehalose and no preservative |
| Gene | UBR2 |
| Gene Accession No. | Q8IWV8 |
| Gene Alias | 9930021A08Rik; AI462103; AW540746; bA49A4.1; C6orf133; dJ242G1.1; dJ392M17.3; E130209G04Rik; E3 ubiquitin-protein ligase UBR2; Kiaa0349; mKIAA0349; N-recognin-2; RING-type E3 ubiquitin transferase UBR2; si:ch211-239i4.2; ubiquitin ligase E3 alpha-II; ubiquitin protein ligase E3 component n-recognin 2; ubiquitin-protein ligase E3-alpha-2; Ubiquitin-protein ligase E3-alpha-II; Ubr2 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.