Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ UBTD1 Monoclonal Antibody (189CT14.5.4)
GREENER_CHOICE

Catalog No. PIMA537493
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA537493 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA537493 Supplier Invitrogen™ Supplier No. MA537493
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

UBTD1 Monoclonal Antibody for Western Blot

The degradation of many proteins is carried out by the ubiquitin pathway in which proteins are targeted for degradation by covalent conjugation of the polypeptide ubiquitin. This gene encodes a protein that belongs to the ubiquitin family of proteins. The encoded protein is thought to regulate E2 ubiquitin conjugating enzymes belonging to the UBE2D family.
TRUSTED_SUSTAINABILITY

Specifications

Antigen UBTD1
Applications Western Blot
Classification Monoclonal
Clone 189CT14.5.4
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene UBTD1
Gene Accession No. Q9HAC8
Gene Alias BC016129; FLJ11807; ubiquitin domain containing 1; ubiquitin domain-containing protein 1; Ubtd1
Gene Symbols UBTD1
Host Species Mouse
Immunogen UBTD1 recombinant protein.
Purification Method Protein G
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 80019
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.