Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

UBXN4 Polyclonal Antibody, Bethyl Laboratories

Catalog No. 501570915
Encompass_Preferred
Change view
Click to view available options
Quantity:
20 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-157-0915 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-157-0915 Supplier Bethyl Laboratories, Inc Supplier No. A305635AT
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Mouse, Rat, Bovine and Orangutan

UBXD2 is an integral membrane protein of the endoplasmic reticulum (ER) that binds valosin-containing protein and promotes ER-associated protein degradation.

Specifications

Antigen UBXN4
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation TBS with 0.1% BSA and 0.09% sodium azide
Gene UBXN4
Gene Accession No. Q92575
Gene Alias erasin, KIAA0242, UBX domain containing 2, UBX domain-containing 1, UBX domain-containing protein 2, UBXD2, UBXDC1
Gene Symbols UBXN4
Host Species Rabbit
Immunogen Between 458 and 508
Purification Method Antigen affinity chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23190
Target Species Human
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.