Learn More
Dräger™ UCF 1600 Thermal Imaging Camera Accessories
Catalog No. 17987304 Shop All Draeger Safety, Inc. Products
Change view
Click to view available options
Product Type:
Crawling Handle
Desktop Battery Charger
Lithium Ion Battery
Picture/Video Capture Handle
Retractable Lanyard
Shoulder Strap
Tripod
Truck Charger
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
17-987-304 Picture/Video Capture Handle
17-987-307 Truck Charger
17-987-302 Crawling Handle
17-987-314 Retractable Lanyard
17-987-306 Desktop Battery Charger
17-987-313 Shoulder Strap
17-987-315 Tripod
17-987-305 Lithium Ion Battery
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. 17-987-304 Supplier Dräger™ Supplier No. 4057991
Camera delivers excellent image quality under the most extreme conditions
Specifications
Product Type Picture/Video Capture Handle
For Use With (Application) Saves more than 60 min. of video and more than 1000 images
For Use With (Equipment) UCF 1600 and 3200 thermal imaging cameras