Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

UCL 2077, Tocris Bioscience™
SDP

Catalog No. 266110 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
26-611-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 26-611-0 Supplier R&D Systems Supplier No. 2661/10

Item Discontinued Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Slow afterhyperpolarization (sAHP) channel blocker; sub-type selective Kv channel effects

Slow afterhyperpolarization (sAHP) channel blocker; reduces sAHP in hippocampal slice preparations. Displays no effect on Ca2+ currents or the time course of sAHP/sIAHP. Exhibits potent inhibition of KCNQ1 and KCNQ2, but differential effects at other KCNQs. Rescues memory retrieval in dopamine β-hydroxylase knockout mice.

Chemical Identifiers

CAS 918311-87-2
Molecular Formula C25H22N2
Molecular Weight (g/mol) 350.47
MDL Number MFCD00585879
InChI Key PQFNWDHABGBCHB-UHFFFAOYSA-N
Synonym 3-triphenylmethylaminomethyl pyridine, pyridin-3-ylmethyl triphenylmethyl amine, n-trityl-3-pyridinemethanamine, 3-tribenzylaminomethyl pyridine, ucl hplc , solid, 1,1,1-triphenyl-n-pyridin-3-yl methyl methanamine
PubChem CID 24868317
IUPAC Name [(pyridin-3-yl)methyl](triphenylmethyl)amine
SMILES C(NC(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1)C1=CN=CC=C1

Specifications

Quantity 10 mg
Formula Weight 350.46
Percent Purity >99%
Chemical Name or Material UCL 2077
Recommended Storage Store at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.