Learn More
Invitrogen™ UCN3 Monoclonal Antibody (C9), Invitrogen™
Description
Specifications
Specifications
| Antigen | UCN3 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Clone | C9 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 |
| Gene | UCN3 |
| Gene Accession No. | Q969E3 |
| Gene Alias | prepro-urocortin 3; RGD1562213; SCP; SPC; Stresscopin; ucn III; Ucn3; UCNIII; urocortin 3; urocortin 3 (stresscopin); Urocortin II; Urocortin III; Urocortin-3 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.