Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ UCP1/UCP3 Recombinant Rabbit Monoclonal Antibody (0S10O10)
GREENER_CHOICE

Catalog No. PIMA537913
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA537913 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA537913 Supplier Invitrogen™ Supplier No. MA537913
Only null left
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Positive Samples: Mouse skeletal muscle, Mouse fat.

UCP are mitochondrial transporter proteins that create proton leaks across the inner mitochondrial membrane, thus uncoupling oxidative phosphorylation. As a result, energy is dissipated in the form of heat. May play a role in the modulation of tissue respiratory control. Participates in thermogenesis and energy balance.
TRUSTED_SUSTAINABILITY

Specifications

Antigen UCP1/UCP3
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone 0S10O10
Concentration 1.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol, 0.05% BSA and 0.02% sodium azide; pH 7.3
Gene UCP1
Gene Accession No. P04633, P12242, P56499, P56501
Gene Alias AI385626; AI645527; mitochondrial brown fat uncoupling protein 1; Mitochondrial uncoupling protein 3; SLC25A7; SLC25A9; solute carrier family 25 member 7; Solute carrier family 25 member 9; thermogenin; Ucp; UCP 1; UCP 3; Ucp1; UCP3; UCP-3; Ucpa; uncoupling protein 1; uncoupling protein 1 (mitochondrial, proton carrier); uncoupling protein 1, mitochondrial; uncoupling protein 3; uncoupling protein 3 (mitochondrial, proton carrier); Uncoupling protein 3, mitochondrial; uncoupling protein, mitochondrial; Uncp
Gene Symbols UCP1, UCP3
Host Species Rabbit
Immunogen Recombinant protein (or fragment).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22227, 22229, 24860, 25708
Target Species Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.