Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Velp™ Scientific UDK149 Distillation Unit

Automatic distillation for Kjeldahl protein determination, non-protein nitrogen (NPN), volatiles and more. Offers connection to external potentiometric titrators for higher sample throughput, flexibility and premium precision.
Supplier: Velp™ Scientific F30200140
This item is not returnable.
View return policy
Description
- Designed for maximum versatility and reproducible results when determining Kjeldahl nitrogen TKN, proteins, ammoniacal nitrogen, nitric nitrogen (Devarda), TVBN, sulfites, phenols, volatile acids, cyanides, and alcohol content
- Can be used in combination with several models of external potentiometric titrators for higher sample throughput
- Programmable boric acid, water, and sodium hydroxide addition
- Selectable steam generation output level of 10 to 100%
- Auto removal of residues from sample tube
- Unique VELP™ patented titanium condenser designed for efficient thermal exchange and low water consumption
- Distillate temperature is always kept below Kjeldahl threshold value (35°C), without nitrogen loss
- Reduced water consumption
- High resistance material with prolonged lifespan
- VELP™ Technopolymer Splash Head is designed for high throughput
Specifications
230 V | |
Distillation Unit | |
50/60 Hz |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction