Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ UDP-Galactose Transporter, Mouse, Unlabeled, Clone: 1H10.1,

Catalog No. MABC1205MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
MABC1205MI 100 μL Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABC1205MI Supplier MilliporeSigma™ Supplier No. MABC1205
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Apolipoprotein E (Apo E) is a polymorphic, multifunctional protein synthesized by liver, kidney, brain, and other cell types. It has been reported that the genetic inheritance of apolipoprotein isoform E4 is a significant risk factor for late onset Alzheimer's Disease (AD).
TRUSTED_SUSTAINABILITY

Specifications

Antigen UDP-Galactose Transporter
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 1H10.1
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Formulation Purified mouse IgG3κ in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% Sodium Azide.
Gene SLC35A2, CDG2M, UGALT, UGT, UGTL
Gene Accession No. P78381
Host Species Mouse
Immunogen KLH-conjugated linear peptide corresponding to human UDP-Gal transporter C-terminal end sequence.
Purification Method Protein G Purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Apoptosis & Cancer
Primary or Secondary Primary
Gene ID (Entrez) NP_005651
Test Specificity The immunogen sequence is 100% present in spliced isoforms 1 (UGT2) and 3, while being only partially present in spliced isoforms 2 (UGT1), 4, and 5 reported by UniProt (P78381), reactivity toward these latter isoforms has not been confirmed.
Target Species Human
Content And Storage Stable for 1 year at 2°-8°C from date of receipt.
Form Purified
Isotype IgG3 κ
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.