Learn More
Medchemexpress LLC UDP-GlcNAc disodium salt | 91183-98-1 | MFCD00799985 | 99.9% | 651.32 g/mol | C17H25N3Na2O17P2 | 5 MG
Supplier: Medchemexpress LLC HY1121745MG
UDP-GlcNAc disodium salt is the disodium salt of uridine diphosphate N-acetylglucosamine, an activated nucleotide sugar used as the donor substrate for O-GlcNAc transferase (OGT) in enzymatic glycosylation studies. It is supplied as a white to off-white solid with high purity and is intended for biochemical and enzymology research.
- Donor substrate for O-GlcNAc transferase-mediated protein O-GlcNAcylation.
- High purity (99.9%) suitable for analytical and enzymatic assays.
- Supplied as a stable solid for long-term storage at -20°C.
- Soluble in aqueous buffers for in vitro reaction use.
- Available in multiple small-scale sizes for research convenience.