Learn More
Promega UDP-Glo Glycosyltransferase Assay, 400 assays
Catalog No. PRV6962 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
400 Assays
Catalog No. Quantity
PRV6962 400 Assays
1 options
Catalog No. PRV6962 Supplier Promega Supplier No. V6962
The UDP-Glo Glycosyltransferase Assay is a bioluminescent assay for detecting the activity of glycosyltransferases that use UDP-sugars as donor substrates and release UDP as a product.
  • Universal Glycosyltransferase Activity Assay
  • Easy, “add−incubate−read” protocol
  • Detects any glycosyltransferase that uses UDP−sugar as a substrate
  • Ideal for low−activity glycosyltransferases
Specifications
Content And Storage <-65°C
Quantity 400 Assays