Learn More
Medchemexpress LLC UDP-glucosamine (UDP-GlcNAc) disodium | 1355005-51-4 | 99.4% | C15H23N3Na2O16P2 | 10MG

Supplier: Medchemexpress LLC HY13979310MG
UDP-glucosamine disodium is the disodium salt of uridine diphosphate glucosamine (UDP-GlcNAc), a nucleotide sugar substrate used by O-GlcNAc transferase for protein O-GlcNAcylation. It is supplied as a white to off-white solid with high reported purity and strong aqueous solubility, intended for biochemical and enzymatic assays.
- High purity (99.4%) suitable for biochemical assays.
- White to off-white solid for convenient handling and storage.
- Soluble in water (250 mg/mL; ultrasonic may be required) for assay preparation.
- Stable when stored sealed and kept away from moisture; solution stability recommended at low temperatures.
- Functionally active as a substrate for O-GlcNAc transferase in protein modification studies.
- Molecular weight 609.28 g/mol and molecular formula C15H23N3Na2O16P2.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.