Learn More
Grainger UEI TEST INSTRUMENTS Flue Gas Particle Filter, Water Trap
Supplier:  Grainger 6CMNO
Encompass_Preferred
The UEI TEST INSTRUMENTS Flue Gas Particle Filter Water Trap is designed to remove moisture and particulates from flue gas samples. It features a clear bowl to allow visual inspection of collected moisture and has a drain valve to remove collected moisture. The filter element removes particulates down to 10 microns. This accessory connects inline with flue gas analyzers and can help protect sensitive measurement instrumentation from moisture and dirt. It has 3/8 inch compression fittings for easy installation.
Catalog No. NC2823652
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.