Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ UEVLD Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5101949
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5101949 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5101949 Supplier Invitrogen™ Supplier No. PA5101949
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total UEVLD.

Possible negative regulator of polyubiquitination.
TRUSTED_SUSTAINABILITY

Specifications

Antigen UEVLD
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene UEVLD
Gene Accession No. Q8IX04
Gene Alias 8430408E05Rik; Attp; EV and lactate/malate dehydrogenase domain-containing protein; signaling molecule ATTP; ubiquitin-conjugating enzyme E2 variant 3; UEV and lactate/malate dehyrogenase domains; Uev3; UEV-3; UEVLD
Gene Symbols UEVLD
Host Species Rabbit
Immunogen A synthesized peptide derived from human UEVLD(Accession Q8IX04), corresponding to amino acid residues G65-Q115.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55293
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.