Description
Allows for conducting fully automated memory experiments including:
- Assessment of spatial memory
- Basic working memory
- Discriminating working from reference memory
- Evaluating impairments in working memory
Includes a central electronic unit and up to eight modular units that can be configured as:
- Y-maze
- T-maze
- 8-arm Radial Arm Maze
Proprietary sliding doors retract into the maze floor smoothly and silently without disrupting the animal during testing. This sliding mechanism combined with the non-reflective, high contrast maze material provides optimal results with ANY-maze Video Tracking Software (sold separately).
Specifications
Specifications
| For Use With (Application) | Neuroscience |
| Animal | Rat |
| Model | Standard arm with automated door |