Learn More
Stoelting™ Ugo Basile Sociability Apparatus
SDP
Catalog No. 10001065 Shop All Stoelting Co Products
Change view
Click to view available options
Animal:
Mouse
Rat
Model:
Opaque with gray enclosures
With 2 stranger enclosures
With white enclosures
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Animal Model
10-001-065 Mouse With white enclosures
10-001-061 Rat Opaque with gray enclosures
10-001-062 Rat With white enclosures
10-001-063 Mouse With 2 stranger enclosures
10-001-067 Mouse Opaque with gray enclosures
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 10-001-065 Supplier Stoelting™ Supplier No. 60456
Only null left
Add to Cart
Add to Cart
Request a Quote
Stoelting™ Sociability Apparatus is a 3-chambered social test used to study social behaviour in mice. Useful for research on autism, parental behavior, sociability and social memory (social novelty).
Ideal for the investigation of autism-related behaviors:
  • Social approach
  • Resistance to reverse a learned pattern of behavior
Shortly after a period of habituation a mouse's sociability is determined by evaluating the amount of time the test mouse spends approaching a wire cage containing an unfamiliar stranger mouse.
In the second phase of testing a second unfamiliar mouse is introduced into the Sociability Apparatus. The preference for social novelty is then determined by measuring the amount of time the test mouse spends approaching the chamber containing a new, unfamiliar stranger mouse (also restricted to a small wire cage), as compared to time spent in the chamber containing the now-familiar mouse. Resistance to change is measured by two reversal learning tasks.
Testing in the Sociability Apparatus can be completely automated by the ANY-maze software system.
Specifications
For Use With (Application) Neuroscience
Animal Mouse
Model With white enclosures