Learn More
Invitrogen™ UGT1A10 Polyclonal Antibody

Description
The antibody detects endogenous levels of total UGT1A10 protein.
Specifications
Specifications
| Antigen | UGT1A10 |
| Applications | Immunohistochemistry, Western Blot |
| Classification | Polyclonal |
| Concentration | 1.3 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 |
| Gene | UGT1A10 |
| Gene Accession No. | Q6ZQM8, Q9HAW8 |
| Gene Alias | A10'; A13; GNT1; UDP glucuronosyltransferase 1 family, polypeptide A10; UDP glucuronosyltransferase 1 family, polypeptide A7C; UDP glucuronosyltransferase family 1 member A10; UDP glycosyltransferase 1 family polypeptide A13; UDP glycosyltransferase 1 family, polypeptide A10; UDP-glucuronosyltransferase 1-10; UDP-glucuronosyltransferase 1-7C; UDP-glucuronosyltransferase 1A10; UDP-glucuronosyltransferase 1A7C; UDP-glucuronosyltransferase 1-J; UDPGT; UDPGT 1-10; UDPGT 1-7C; UGT1; UGT1*7C; UGT1.10; UGT1.7C; UGT1-07C; UGT1-10; Ugt1a10; Ugt1a13; Ugt1a7c; UGT1J; UGT-1J |
| Gene Symbols | UGT1A10, Ugt1a7c |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.