Learn More
Strem, An Ascensus Company CAS# 1072413-89-8. 500mg. Zirconium 1,4-dicarboxybenzene MOF (UiO-66).
SDP
Supplier:  Strem, An Ascensus Company 401105500MG
Encompass
CAS# 1072413-89-8. 500mg. Zirconium 1,4-dicarboxybenzene MOF (UiO-66). . Molecular Weight: 1664.06. Molecular Formula: C48H28O32Zr6. Color/form: white pwdr. Strem# 40-1105. http://www.strem.com/catalog/v/40-1105/
Catalog No. 50-204-5303
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.