Learn More
Medchemexpress LLC UK-101 | 1000313-40-5 | 99.6% | 484.74 g/mol | 50 MG
SDP
Supplier:  Medchemexpress LLC HY11903750MG
Encompass_Preferred
UK-101 is a potent, selective inhibitor of the immunoproteasome β1i (LMP2) subunit, commonly used in proteasome biology and cancer research to study apoptosis and proteasome-mediated pathways.
  • Potent LMP2 inhibition with an IC50 of about 104 nM.
  • Demonstrates high selectivity versus related proteasome subunits for target-specific studies.
  • Induces cell apoptosis, useful in cancer biology assays.
  • High purity (≈99.6%) for reproducible experimental results.
  • Soluble in DMSO (≈7.3 mg/mL) for easy stock preparation.
  • Stable as a powder under low-temperature storage for long-term use.
  • Provided in a small research pack suitable for laboratory assays.
Catalog No. 50-002-26984