Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

UK 14,304 tartrate, Tocris Bioscience™
GREENER_CHOICESDP

Catalog No. 246610 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
24-661-0 10 mg
24-665-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 24-661-0 Supplier Novus Biologicals Supplier No. 2466/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

α2 agonist. Water-soluble form of UK 14,304 (Cat. No. 0425)

Water-soluble form of UK 14,304 (Cat. No. 0425). Full α2 adrenergic agonist. Centrally active following systemic administration in vivo.

Chemical Identifiers

CAS 70359-46-5
Molecular Formula C15H16BrN5O6
Molecular Weight (g/mol) 442.226
InChI Key QZHBYNSSDLTCRG-YKWZRPGSSA-N
Synonym brimonidine tartrate, --tartaric acid; brimonidine
PubChem CID 72376318
IUPAC Name 5-bromo-N-(4,5-dihydro-1H-imidazol-2-yl)quinoxalin-6-amine;(2R,3S)-2,3-dihydroxybutanedioic acid
SMILES C1CN=C(N1)NC2=C(C3=NC=CN=C3C=C2)Br.C(C(C(=O)O)O)(C(=O)O)O

Specifications

Quantity 10 mg
Formula Weight 442.23
Percent Purity >99%
Chemical Name or Material UK 14, 304 tartrate
Recommended Storage Desiccate at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.