Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ULBP2/ULBP5/ULBP6 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA547118 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA547118 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA547118 Supplier Invitrogen™ Supplier No. PA547118
Add to Cart
Edge
Add to Cart

Goat Polyclonal Antibody

In direct ELISAs and Western blots, less than 2% cross-reactivity with recombinant human (rh) ULBP-1, rhULBP-3, and rhULBP-4 is observed. Reconstitute at 0.2 mg/mL in sterile PBS. Endoxin level is <0.10 EU per 1 μg of the antibody by the LAL method.

TRUSTED_SUSTAINABILITY

Specifications

Antigen ULBP2/ULBP5/ULBP6
Applications Immunohistochemistry (Paraffin), Neutralization, Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 5% trehalose and No Preservative
Gene RAET1G
Gene Accession No. Q6H3X3, Q9BZM5
Gene Alias ALCAN-alpha; N2DL2; N2DL-2; NKG2D ligand 2; NKG2DL2; RAET1G; RAET1H; retinoic acid early transcript 1G; retinoic acid early transcript 1G protein; Retinoic acid early transcript 1H; UL16 binding protein 2; UL-16 binding protein 5; UL16-binding protein 2; ULBP; ULBP2; ULBP5; UNQ463/PRO791
Gene Symbols RAET1G, ULBP2
Host Species Goat
Immunogen Mouse myeloma cell line NS0-derived recombinant human ULBP-2 Gly26-Ser217.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 353091, 80328
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.