Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ULBP3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA547148
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA547148 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA547148 Supplier Invitrogen™ Supplier No. PA547148
Only null left
Add to Cart
Add to Cart

Goat Polyclonal Antibody

Approximately 5% cross-reactivity with recombinant human (rh) ULBP-2 is observed and less than 1% cross-reactivity with rhULBP-1 is observed. Reconstitute at 0.2 mg/mL in sterile PBS. Endoxin level is <0.10 EU per 1 μg of the antibody by the LAL method.

UL16-binding protein 3 (ULBP3) is an extended member of the MHC class I superfamily. It does not contain the alpha 3 domain and lacks a transmembrane domain. ULBP3 binds and activates the KLRK1/NKG2D receptor and mediates natural killer cell cytotoxicity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ULBP3
Applications Neutralization, Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 5% trehalose and No Preservative
Gene ULBP3
Gene Accession No. Q9BZM4
Gene Alias ALCAN-gamma; N2DL3; N2DL-3; NKG2D ligand 3; NKG2D ligand3; NKG2DL3; RAET1N; Retinoic acid early transcript 1N; UL16 binding protein 3; UL16-binding protein 3; ULBP; ULBP 3; ULBP3
Gene Symbols ULBP3
Host Species Goat
Immunogen Mouse myeloma cell line NS0-derived recombinant human ULBP-3 Gly27-Pro216.
Purification Method Antigen Affinity Chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79465
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.