Learn More
Biolegend Ultra-LEAF™ Purified anti-DYKDDDDK Tag
Supplier:  Biolegend 637328
Encompass_Preferred
Ultra-LEAF™ Purified anti-DYKDDDDK Tag [L5]; Isotype: Rat IgG2a, λ; Reactivity: DYKDDDDK tag epitope; Apps: WB, IP, ICC, ELISA, FC, Purification; Size: 1 mg
Catalog No. 50-208-6872
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.