Learn More
Biolegend Ultra-LEAF™ Purified anti-mouse CD254 (TRANCE, RANKL)
Supplier:  Biolegend 510009
Encompass_Preferred
Ultra-LEAF™ Purified anti-mouse CD254 (TRANCE, RANKL) [IK22/5]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: FC, IP, WB, Block; Size: 100 ug
Catalog No. 50-208-5807
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.