Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ulipristal Acetate 98.0+%, TCI America™
SDP

Catalog No. U0102250MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
50 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
U0102250MG 250 mg
U010250MG 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. U0102250MG Supplier TCI America Supplier No. U0102250MG
Add to Cart
Add to Cart

Chemical Identifiers

CAS 126784-99-4
Molecular Formula C30H37NO4
Molecular Weight (g/mol) 475.629
MDL Number MFCD00899035
InChI Key OOLLAFOLCSJHRE-ZHAKMVSLSA-N
Synonym (11beta)-17-(Acetyloxy)-11-[4-(dimethylamino)phenyl]-19-norpregna-4,9-diene-3,20-dione, CDB-2914
PubChem CID 130904
ChEBI CHEBI:71025
IUPAC Name [(8S,11R,13S,14S,17R)-17-acetyl-11-[4-(dimethylamino)phenyl]-13-methyl-3-oxo-1,2,6,7,8,11,12,14,15,16-decahydrocyclopenta[a]phenanthren-17-yl] acetate
SMILES CC(=O)C1(CCC2C1(CC(C3=C4CCC(=O)C=C4CCC23)C5=CC=C(C=C5)N(C)C)C)OC(=O)C

Specifications

Melting Point 186°C
Color Yellow
Quantity 250 mg
Formula Weight 475.63
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Ulipristal Acetate
TSCA No
Recommended Storage Freezer (-20 °C)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.