Learn More
Invitrogen™ ULK1 (denatured) Polyclonal Antibody

Description
ULK1 (denatured) Polyclonal Antibody for Western Blot, IHC (P), Flow
Specifications
Specifications
| Antigen | ULK1 (denatured) |
| Applications | Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.09% sodium azide; pH 7.4 |
| Gene | ULK1 |
| Gene Accession No. | O70405, O75385 |
| Gene Alias | APG1; ATG 1; ATG1; ATG1 autophagy related 1 homolog; ATG1A; AU041434; Autophagy-related protein 1 homolog; C. elegans; EC:2.7.11.1; FLJ38455; hATG1; KIAA0722; mKIAA0722; Serine/threonine-protein kinase ULK1; serine/threonine-protein kinase Unc51.1; ULK 1; Ulk1; UNC 51; UNC51; unc-51 like autophagy activating kinase 1; unc-51 like kinase 1; Unc51.1; unc-51-like kinase 1 |
| Gene Symbols | ULK1 |
| Host Species | Rabbit |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.