Learn More
Thermco Products ULT Calibrated Smart Probe
Catalog No. 04500323
Change view
Click to view available options
For Use With (Equipment):
Data Loggers
Catalog No. For Use With (Equipment)
04-500-323 Data Loggers
1 options
Catalog No. 04-500-323 Supplier Thermco Products Supplier No. LTCP30ULF
Add to Cart
Add to Cart
These precise and reliable probes are compatible with UTREL30-16 and UTREL30-WiFi dual-channel loggers for ultra-low temperature monitoring.
  • Measure temperatures from -90°C to 40°C (-130°F to 104°F), making them ideal for cryogenic applications.
  • Designed for seamless integration, allowing probes to be swapped and calibrated independently.
  • Hot-swappable, plug-and-play design minimizes downtime and eliminates the need to return or relocate the logger for calibration.
  • Expired probes can be sent to a calibration lab while logging continues uninterrupted.
  • Ensures continuous, accurate temperature monitoring for critical applications such as vaccine storage.
  • Supports compliance and operational efficiency through reliable, continuous monitoring.
Specifications
Type Probe
For Use With (Equipment) Data Loggers