Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eisco™ Ultimate Lab Starter Kit
SCI_ED

Catalog No. S487653 Shop All Eisco Products
Change view
Click to view available options
Class Size:
25 Students
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Class Size
S487653 25 Students
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. S487653 Supplier Eisco™ Supplier No. ULTLABKIT
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This ultimate laboratory starter set includes beakers, cylinders, test tubes, burette, clips, clamps, tongs, racks, base, rod, and other essential lab accessories

    Includes:
  • (2) 100 mL Graduated clyinders - polypropylene, hexagonal base, class B tolerance
  • (2)500 mL Beakers, polypropylene, 10 mL graduations
  • (1) 50 mL Acrylic burette with PTFE stopcock
  • 250 mL Filter flask, borosilicate glass, 50 mL graduations
  • (48) 20 mL Test tubes, borosilicate glass, ungraduated
  • (1) Drying basket, 6 x 4.25 x 4.5 in., polypropylene
  • Retort base, 8 x 5 in., powder coated alloy, rubber lined bottom
  • Metal rod, 12 mm diameter with 10 mm threading
  • Bosshead, powder coated
  • Lab clamp, powder coated
  • Z-Shaped test tube/instrument rack with space for up to 17 test tube or pieces of equipment, powder coated and chemical resistant
  • Tripod with wire mesh surface plate
  • Ring clamp with bosshead
  • Test tube brush
  • Flask tongs
  • Spatula with spoon end and flat end
  • Crucible tongs
  • Test tube tongs

Specifications

Grade 5 to 15
Age 10 to 20
Material Stainless Steel, Steel, Polypropylene, Borosilicate Glass
Weight (English) 18 lb.
Weight (Metric) 8.16 kg
Class Size 25 Students
Product Type Labware
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.