Learn More
MINNEBIO LLC Ultra-Active Nuclease (Serratia marcescens Nuclease, EC 3.1.30.2) Benzonase Alternative Purity 98% 500 kU.
Supplier:  MINNEBIO LLC MBPN01500KU
Encompass_Preferred
Ultra-Active Nuclease, derived from Serratia marcescens, is a broad-spectrum endonuclease capable of non-specific cleavage within nucleotide chains, degrading all types of DNA and RNA (double-stranded, single-stranded, linear, and circular). The primary application of this product is the removal of residual nucleic acids from cell lysates, effectively and accurately for various bio-analyses or to streamline downstream processes and improve product purity. The advantages of Ultra-Active Nuclease include: (1) Fast and Efficient: Cleave nucleic acids rapidly, exhibiting a catalytic rate nearly 15-fold faster than that of deoxyribonuclease (DNase). (2) High Tolerance: High tolerance against the reduction and denaturation of harsh conditions, enabling efficient cleavage of nucleic acids under a wide range of conditions. (3) Resistance to Inhibition: Resistant to DNase and RNase inhibitors, ensuring its effectiveness even when other nucleases become inactive.
Catalog No. NC2590922