Learn More
Genscript Corporation GenCRISPR™ Ultra NLS-Cas9-Research
Supplier:  Genscript Corporation Z036211
Encompass
GenCRISPR™ Ultra Cas9 is produced by expression in an E. coli strain carrying a plasmid encoding the Cas9 gene from Streptococcus pyogenes with an N-terminal nuclear localization signal (NLS).
Catalog No. NC2002296
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.