Learn More
Azure Biosystems Ultra Clear Optical Sealing Sheet
SDP
Catalog No. AIQ325AZ Shop All Azure Biosystems Products
Change view
Click to view available options
Adhesive:
Pressure Sensitive
Catalog No. Adhesive
AIQ325AZ Pressure Sensitive
1 options
Catalog No. AIQ325AZ Supplier Azure Biosystems Supplier No. AIQ325
Add to Cart
Add to Cart
Optical, non-piercable seal that can be removed after the qPCR run. Pack of 25
  • Self-adhesive PCR plate sealing sheets that are reliable and used to easily seal 96-well plates for qPCR or PCR applications. They provide non-pierceable protection against evaporation for high efficiency PCR. The sheets are easily removed after the run.
  • Ultra clear
  • Optical
  • Self sealing
  • Pack of 25
Specifications
Color Clear
Material Polyolefin
Sterility Non-sterile
Format Sheet
Pre-slit No
Product Type Sealing 96 well
Adhesive Pressure Sensitive
Thickness 0.75 mm
Pierceable No
Temperature Range (Metric) 0 to 120°C