Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MSA™ Ultra Elite™ Pressure-Demand Full Facepieces

Catalog No. 19815575 Shop All MSA Products
Change view
Click to view available options
Size:
Medium
Large
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-815-575 Medium
19-815-576 Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19-815-575 Supplier MSA™ Supplier No. 491522
Add to Cart
Add to Cart

Pressure-demand full facepieces feature nosecup and E-Z don head harness.

  • State-of-the-art mask provides comfort and protection
  • Ergonomic design
  • Distortion-free lens allows wide field-of-view
  • 5-point harness allows easy on/off
  • Unique airflow prevents lens misting
  • Face blank has a single-sealing flange, silicone
  • Speech diaphragm makes communication easy to discern

Specifications

Color Black
Product Type Full Facepiece Respirator
For Use With (Equipment) SAR, SCBA
Includes Nosecup, E-Z don head harness
Material Silicone
Size Medium
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.