Learn More
Scientific Instrument Services Inc Edwards ULTRAGRADE™ 19 Oil 1L
Supplier: Scientific Instrument Services Inc H11025015
This item has a minimum qty of 2 per supplier requirements.
Edwards offers a range of new oils, ULTRAGRADE™ Performance 15, 19, 20 & 70, which result from a new process. Most manufacturers use a solvent refining process, whereas these oils are produced by a unique hydrotreating process. This process eliminates nitrogen, sulphur & oxygen & the aromatic hydrocarbons, leaving a clear base stock essentially free of impurities. When combined w/ additives, the ULTRAGRADE™ Performance oils give improved vacuum performance, thermal & oxidation stability & rust inhibition. The anti-oxidants included in the oils enable the pump to withstand high operating temps without oil degradation, so extending intervals between oil changes. The properties of the oils allow them to be used in a wide range of applications. New ULTRAGRADE™ Performance oils are the best choice for large pumps used in heavy-duty, cycling applications.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.