Learn More
Restek™ Ultra-High Pressure Sample Cylinder Valves
Large, durable, Kel-F seat ensures leak-free operation.
Supplier: Restek™ 26314
Description
Alta-Robbins unique valve design incorporates a fully contained soft seat that provides durability and longer lifetime. Tight shut-off is easily achieved with very low torque, yet the valve is rugged enough to withstand overtightening.
- Multiple valve configurations, including dip tube and rupture disks
- Temperature range: -40° to +120°C
- Available stainless-steel or sulfinert-treated
Dip Tube
Provides a headspace above liquefied gases so that, should expansion occur with an increase in temperature, the pressure is not dramatically increased. The dip tube is welded directly to the male inlet of the valve and cut to a length of up to 5.25 in.
Rupture Disks
Function to protect sample cylinders from overpressurization by venting to the atmosphere. The pressure rating on the rupture disk should always be lower than the cylinder.
Specifications
Stainless Steel | |
0.25 in. Male NPTx 0.25 in. Male NPT w/2850 psi rupture disc; UHP | |
-40°C to +120°C | |
-40°F to +248°F |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.