Learn More
HARDVARD BIOSCIENCE INC DBA BIOCHROM Ultra Ninhydrin Kit, Colorimetric Derivatization of Amino Acids, Volume: 2 L, Compatible with Biochrom 30+ Series Analyzers, 90 runs with Protein Hydrolysate, 45 runs with Physiological Fluids
Supplier:  HARDVARD BIOSCIENCE INC DBA BIOCHROM 80600310
Encompass_Preferred
Ultra Ninhydrin Kit, Colorimetric Derivatization of Amino Acids, Volume: 2 L, Compatible with Biochrom 30+ Series Analyzers, 90 runs with Protein Hydrolysate, 45 runs with Physiological Fluids
Catalog No. NC2125670